Order Entry
Belgium
Orders LinkContactUsLinkComponent
Transfer capillaries

Transfer capillaries

80 mm, 1,55 mm, 1,15 mm
  HIRS9201580
 :   HIRSCHMANN
Transfer capillaries
  HIRS9201580
Transfer capillaries
Transfer capillaries

80 mm, 1,55 mm, 1,15 mm

 
67,50 €
   
1 * 1.000 ST
 

Clear glass, both end open.

 
  • Ext.Ø:
    1,55 mm
  • Ø int.:
    1,15 mm
  • Length:
    80 mm
  • Seq:
    18