Order Entry
Belgium
Orders LinkContactUsLinkComponent
Volumetric flasks, class B

Volumetric flasks, class B

50 ml, 10/19, 150 mm
Catalog # HIRS7560175
Supplier:   HIRSCHMANN
Volumetric flasks, class B
Catalog # HIRS7560175
Supplier:  HIRSCHMANN
Volumetric flasks, class B
Volumetric flasks, class B

50 ml, 10/19, 150 mm

Price
34,50 €
Log in for availability
1 * 2 ST
Product Details & Documents

PMP, translucent, with PP NS-stopper.

Specifications
  • NS:
    10/19
  • Capacity:
    50 ml
  • Height:
    150 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.