Order Entry
Belgium
Orders LinkContactUsLinkComponent
Burettes, Pellet pattern, without intermediate stopcock, Schellbach

Burettes, Pellet pattern, without intermediate stopcock, Schellbach

25 ml, 0,05 ml, 500 mm, 0,045 ± ml
Catalog # HIRS344017060
Supplier:   HIRSCHMANN
Burettes, Pellet pattern, without intermediate stopcock, Schellbach
Catalog # HIRS344017060
Supplier:  HIRSCHMANN
Burettes, Pellet pattern, without intermediate stopcock, Schellbach
Burettes, Pellet pattern, without intermediate stopcock, Schellbach

25 ml, 0,05 ml, 500 mm, 0,045 ± ml

Price
254,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Automatic burette, Pellet, class AS, DURAN® with blue graduation.

  • Lateral stopcock with glass key or PTFE spindle
  • Schellbach stripes for enhanced legibility
  • Certified conformity, with dated batch identification

 : Compliance with DIN EN ISO 385.

Specifications
  • Capacity:
    25 ml
  • Division:
    0,05 ml
  • Tolerance:
    0,045 ± ml
  • Scale length:
    500 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.