Order Entry
Belgium
Orders LinkContactUsLinkComponent
Demeter pipettes

Demeter pipettes

1,0/2,0/2,1/2,2 ml
Catalog # 612-4803
Supplier:   HIRSCHMANN
Demeter pipettes
Catalog # 612-4803
Supplier:  HIRSCHMANN
Demeter pipettes
Demeter pipettes

1,0/2,0/2,1/2,2 ml

Price
13,30 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

AR-Glas®.

  • Calibrated to deliver 'Ex'
  • Blue graduation
  • Dilution pipettes with two marks at one and one,1 ml or with four marks at 1,0/2,0/2,1/2,2 ml

Length: 250 mm ±5 mm

Length: 250 mm ±5 mm

Certifications: DIN 12838 and DIN 12839

Spécifications
  • Capacity:
    1,0/2,0/2,1/2,2 ml