Order Entry
Belgium
Orders LinkContactUsLinkComponent
HPLC columns, ZORBAX® Eclipse XDB

HPLC columns, ZORBAX® Eclipse XDB

Rapid resolution cartridge, Guard cartridge, Eclipse XDB-C18, 3,5 µm, 2,10 mm, 15 mm
Catalog # HEWL975700-932
Supplier:   Agilent
HPLC columns, ZORBAX® Eclipse XDB
Catalog # HEWL975700-932
Supplier:  Agilent
HPLC columns, ZORBAX® Eclipse XDB
HPLC columns, ZORBAX® Eclipse XDB

Rapid resolution cartridge, Guard cartridge, Eclipse XDB-C18, 3,5 µm, 2,10 mm, 15 mm

Price
769,00 €
Log in for availability
1 * 3 ST
Product Details & Documents

Good peak shape for basic, acidic and neutral compounds.

  • High performance over a wide pH range (2 to 9)
  • Long lifetime with extra dens bonding and double endcapping
Specifications
  • Column Format:
    Guard cartridge
  • Ø int.:
    2,10 mm
  • Particle Size:
    3,5 µm
  • Description:
    Rapid resolution cartridge
  • Length:
    15 mm
  • Seq:
    110
  • Phase:
    Eclipse XDB-C18