Order Entry
Belgium
Orders LinkContactUsLinkComponent
ICP multi-element standards

ICP multi-element standards

Multi-element calibration Standard-3, 100 ml, (10 elements in diluted 10% hydrochloric acid and 1% nitric acid) Sb, Au, Hf, Ir, Pd, Pt, Rh, Ru, Te, Sn, 10 mg/L, 10% HCl, 1
Catalog # HEWL8500-6948
Supplier:   Agilent
ICP multi-element standards
Catalog # HEWL8500-6948
Supplier:  Agilent
CAS Number:  
ICP multi-element standards
ICP multi-element standards

Multi-element calibration Standard-3, 100 ml, (10 elements in diluted 10% hydrochloric acid and 1% nitric acid) Sb, Au, Hf, Ir, Pd, Pt, Rh, Ru, Te, Sn, 10 mg/L, 10% HCl, 1

CAS Number:  
Price
561,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Documents

Specifications

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    10% HCl
  • Pk:
    1
  • Description:
    Multi-element calibration Standard-3, 100 ml, (10 elements in diluted 10% hydrochloric acid and 1% nitric acid) Sb, Au, Hf, Ir, Pd, Pt, Rh, Ru, Te, Sn

Specifications