Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
ICP multi-element standards

ICP multi-element standards

Multi-element calibration Standard-1, 100 ml, (17 elements in diluted nitric acid) Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb, 10 mg/L, 5% HNO₃, 1
Catalog # HEWL8500-6944
Supplier:   Agilent
ICP multi-element standards
Catalog # HEWL8500-6944
Supplier:  Agilent
CAS Number:  
ICP multi-element standards
ICP multi-element standards

Multi-element calibration Standard-1, 100 ml, (17 elements in diluted nitric acid) Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb, 10 mg/L, 5% HNO₃, 1

CAS Number:  
Price
291,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Documents

Specifications

Specifications

  • Concentration:
    10 mg/L
  • Matrice:
    5% HNO₃
  • Cdt:
    1
  • Désignation:
    Multi-element calibration Standard-1, 100 ml, (17 elements in diluted nitric acid) Ce, Dy, Er, Eu, Gd, Ho, La, Lu, Nd, Pr, Sc, Sm, Tb, Th, Tm, Y, Yb

Specifications