Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

ICP multi-element standards

Multi-element calibration standard-2A, 100 ml, (27 elements in diluted nitric acid) Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn, 10 mg/L, 5% HNO₃, 1
N° de catalogue HEWL8500-6940
Fournisseur:   Agilent
ICP multi-element standards
N° de catalogue HEWL8500-6940
Fournisseur:  Agilent
Numéro CAS:  
ICP multi-element standards
ICP multi-element standards

Multi-element calibration standard-2A, 100 ml, (27 elements in diluted nitric acid) Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn, 10 mg/L, 5% HNO₃, 1

Numéro CAS:  
Prix
704,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

 

Specifications

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    5% HNO₃
  • Pk:
    1
  • Description:
    Multi-element calibration standard-2A, 100 ml, (27 elements in diluted nitric acid) Ag, Al, As, Ba, Be, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn

Specifications