Order Entry
Belgium
Orders LinkContactUsLinkComponent
Organic acids solution kit

Organic acids solution kit

Buffer, organic acid, 250 ml, pH 5,6, 250 mL
Catalog # HEWL8500-6785
Supplier:   Agilent
Organic acids solution kit
Catalog # HEWL8500-6785
Supplier:  Agilent
CAS Number:  
Organic acids solution kit

Buffer, organic acid, 250 ml, pH 5,6, 250 mL

CAS Number:  
Price
188,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

The Organic Acids Solutions Kit contains capillaries, buffers, an organic acids standard and instructions complete with a sample electropherogram to help you begin organic acids analyses immediately.

  • Ideal for the analysis of short alkyl-chain carboxylic acids
  • Analyze food applications, including beer, wine and juice
  • Use for chemical applications, including raw materials and drug formulations

Employing an indirect UV detection agent optimized for organic acids, the methodology is simple, sensitive, and provides accurate quantitative analysis. Suited for the analysis of organic acids in a wide range of matrices, it is especially useful for determination of organic acids in beverages, and food.

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Buffer, organic acid, 250 ml, pH 5,6

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.