Orders LinkContactUsLinkComponent
Inlet septa
Inlet septa
Septa, long-life, non-stick, 11 mm
Catalog # HEWL5183-4761
Supplier:   Agilent
Inlet septa
Catalog # HEWL5183-4761
Supplier:  Agilent
Inlet septa
Inlet septa
Septa, long-life, non-stick, 11 mm
Price
139,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
These septa are available for a variety of applications and have different upper temperature limits. The lower temperature septa are usually softer, seal better and can withstand more punctures (injections) than their high temperature counterparts.
  • Made up low bleed silicone rubber formulations
  • Efficient by using sharp syringe needles
  • Septa should be replaced to avoid leaks and sample loss
  • To be used with recommended temperature range
Specifications
  • Ø:
    11 mm
  • Description:
    Septa, long-life, non-stick