Inlet liners
Straight, direct inlet liner, non-deactivated (for gas samples headspace or purge & trap), 140 μl, 1,5 mm
Catalog # HEWL5183-4709
Supplier: Agilent
Jump to:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Specifications
- Ø int.:1,5 mm
- Description:Straight, direct inlet liner, non-deactivated (for gas samples headspace or purge & trap), 140 μl
- Seq:48
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.