Orders LinkContactUsLinkComponent
Inlet liners
Inlet liners
Splitless, single taper liner with glass wool, deactivated, 900 μl, 4 mm
Catalog # HEWL5183-4693
Supplier:   Agilent
Inlet liners
Catalog # HEWL5183-4693
Supplier:  Agilent
Inlet liners
Inlet liners
Splitless, single taper liner with glass wool, deactivated, 900 μl, 4 mm
Price
192,00 €
Log in for availability
1 * 5 ST
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
  • Consistent high level of inertness
Specifications
  • Ø int.:
    4 mm
  • Description:
    Splitless, single taper liner with glass wool, deactivated, 900 μl
  • Seq:
    33