Inlet liners
Split, single taper liner with glass wool, deactivated, low pressure drop, 870 μl, 4 mm
N° de catalogue HEWL5183-4647
Fournisseur: Agilent
Aller à :
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Spécifications
- Ø int.:4 mm
- Description:Split, single taper liner with glass wool, deactivated, low pressure drop, 870 μl
- Seq:18