Inlet liners
PTV liner, multi baffle, deactivated, 150 µl, 1,8 mm
Catalog # HEWL5183-2037
Supplier: Agilent
Jump to:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Specifications
- Ø int.:1,8 mm
- Description:PTV liner, multi baffle, deactivated, 150 µl
- Seq:59