Orders LinkContactUsLinkComponent
Inlet septa
Inlet septa
Microseal high pressure nut
Catalog # HEWL5182-3445
Supplier:   Agilent
Inlet septa
Catalog # HEWL5182-3445
Supplier:  Agilent
Inlet septa
Inlet septa
Microseal high pressure nut
Price
253,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These septa are available for a variety of applications and have different upper temperature limits. The lower temperature septa are usually softer, seal better and can withstand more punctures (injections) than their high temperature counterparts.
  • Made up low bleed silicone rubber formulations
  • Efficient by using sharp syringe needles
  • Septa should be replaced to avoid leaks and sample loss
  • To be used with recommended temperature range
Specifications
  • Description:
    Microseal high pressure nut