Capillary column splitter kits
Fixed inlet splitter kit, 0,53 mm
Catalog # HEWL5181-3392
Supplier: Agilent
Jump to:
Product Details & Documents
These splitters are used for splitting an injected sample from a single injector into two columns and on to two detectors.
The dimensions of both the columns, length and internal diameter (ID), should be the same so that the vaporized sample will get spilt into almost equal amount on to two columns.
Delivery information: Kit which comes with transfer line, splitter union and necessary ferrules.
Specifications
- Ø int.:0,53 mm
- Description:Fixed inlet splitter kit
- Seq:03