Order Entry
Belgium
Orders LinkContactUsLinkComponent
Particle traps for use with PLOT columns

Particle traps for use with PLOT columns

Particle trap for use with PLOT columns, Int.Ø×L: 0,53 mm × 2,5 m
Catalog # HEWL5181-3352
Supplier:   Agilent
Particle traps for use with PLOT columns
Catalog # HEWL5181-3352
Supplier:  Agilent
Particle traps for use with PLOT columns

Particle trap for use with PLOT columns, Int.Ø×L: 0,53 mm × 2,5 m

Price
353,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Though highly stabilised, it is impossible to guarantee that no particles will dislodge from the column wall. When used in valve-switching applications, the use of a particle trap can prevent scarring of the rotors.

Specifications
  • Description:
    Particle trap for use with PLOT columns, Int.Ø×L: 0,53 mm × 2,5 m
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.