Inlet liners
Splitless, single taper liner with glass wool, deactivated, 900 μl, 4 mm
Catalog # HEWL5062-3587
Supplier: Agilent
Jump to:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Specifications
- Ø int.:4 mm
- Description:Splitless, single taper liner with glass wool, deactivated, 900 μl
- Seq:32