Macro cuvettes
Macro, 700 µl, 2 mm, Quartz
HEWL5061-3385
: Agilent
Cells designed for the 8453 UV/Visible spectrophotometers for optical excellence.
- PTFE lid or stopper
- Path length:2 mm
- Type:Macro
- Volume:700 µl
- Window material:Quartz
- Seq:004
Accessories
- Description
- Description
- Description
- Description
- Description
- Description