Order Entry
Belgium
Orders LinkContactUsLinkComponent
Solvent filter/degasser

Solvent filter/degasser

RC filter membrane, 0,45 μm, 47 mm
Catalog # HEWL3150-0576
Supplier:   Agilent
Solvent filter/degasser
Catalog # HEWL3150-0576
Supplier:  Agilent
Solvent filter/degasser
Solvent filter/degasser

RC filter membrane, 0,45 μm, 47 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

It prevents the formation of spurious peaks within the detector due to solvent outgassing at the low-pressure end of the chromatograph.

  • Degasses eluents as particulates are removed
  • Increases solvent inlet lifetime
  • Eliminates pump downtime caused by air locks and particulates in check valves
  • Decreases piston wear, while increasing column life
Specifications
  • Ø:
    47 mm
  • Pore size:
    0,45 μm
  • Description:
    RC filter membrane
  • Seq:
    03
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.