Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Solvent filter/degasser

PTFE filter membrane, 0,45 μm, 47 mm
  HEWL3150-0509
 :   Agilent
Solvent filter/degasser
  HEWL3150-0509
 :  Agilent
Solvent filter/degasser
Solvent filter/degasser

PTFE filter membrane, 0,45 μm, 47 mm

 
 
   
1 * 10 ST
 

It prevents the formation of spurious peaks within the detector due to solvent outgassing at the low-pressure end of the chromatograph.

  • Degasses eluents as particulates are removed
  • Increases solvent inlet lifetime
  • Eliminates pump downtime caused by air locks and particulates in check valves
  • Decreases piston wear, while increasing column life
 
  • Ø:
    47 mm
  • Pore size:
    0,45 μm
  • Description:
    PTFE filter membrane
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.