Orders LinkContactUsLinkComponent
Micro pipettes, disposable, with ring mark
Micro pipettes, disposable, with ring mark
25 µl, 2 × white, One mark
Catalog # HECH40555015
Micro pipettes, disposable, with ring mark
Catalog # HECH40555015
Micro pipettes, disposable, with ring mark
Micro pipettes, disposable, with ring mark
25 µl, 2 × white, One mark
Price
34,10 €
Log in for availability
1 * 250 ST
Product Details & Documents
Disposable micro pipettes with ring mark.
  • Calibrated to contain 'In'
  • Colour-coded
Certifications: DIN ISO 7550
Specifications
  • Colour code:
    2 × white
  • Type:
    One mark
  • Capacity:
    25 µl
  • Seq:
    02