Orders LinkContactUsLinkComponent
Staining jars
Staining jars
Staining dishes acc. to Schiefferdecker, for up to 10 slides
Catalog # 631-9313
Staining jars
Catalog # 631-9313
Staining jars
Staining jars
Staining dishes acc. to Schiefferdecker, for up to 10 slides
Price
26,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Soda-lime glass.
  • Three versions
  • For 16 or 10 slides
  • Acc. to Hellendahl or Schiefferdecker
Suitable for standard microscope slides (76×26 mm).
Specifications
  • Description:
    Staining dishes acc. to Schiefferdecker, for up to 10 slides