Orders LinkContactUsLinkComponent
Staining jars
Staining jars
Rectangular staining jar with cover, 40×20×85 mm, - mm, - mm
Catalog # 631-9314
Staining jars
Catalog # 631-9314
Staining jars
Staining jars
Rectangular staining jar with cover, 40×20×85 mm, - mm, - mm
Price
12,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Soda-lime glass.
These staining jars have an overlapping cover.
Specifications
  • Ø:
    - mm
  • Description:
    Rectangular staining jar with cover
  • Height:
    - mm
  • W×D×H:
    40×20×85 mm