Orders LinkContactUsLinkComponent
Staining racks
Staining racks
Staining rack, extendable, Plexiglas® with PVC tray and clamps
Catalog # 631-9156
Staining racks
Catalog # 631-9156
Staining racks
Staining racks
Staining rack, extendable, Plexiglas® with PVC tray and clamps
Price
109,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Staining rack, Plexiglas®, in the form of an extendable bridge, that can be fixed in the horizontal or at an inclined position. Suitable for trays or dishes of 380 to 620 mm in length.
Specifications
  • Description:
    Staining rack, extendable, Plexiglas® with PVC tray and clamps