Orders LinkContactUsLinkComponent
Cover glasses
Cover glasses
#1, 13 mm
Catalog # 631-9508
Cover glasses
Catalog # 631-9508
Cover glasses
Cover glasses
#1, 13 mm
Price
132,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Borosilicate glass highly resistant cover glasses with precise and faultless cut.
Ordering information: Available in different diameters.
Specifications
  • Ø:
    13 mm
  • No.:
    #1
  • Seq:
    06