Cover glasses
#1, 10 mm
Catalog # HECH41001110
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Borosilicate glass highly resistant cover glasses with precise and faultless cut.
Ordering information: Available in different diameters.
Specifications
- Ø:10 mm
- No.:#1
- Seq:04