Order Entry
Belgium
Orders LinkContactUsLinkComponent
Dissecting needles

Dissecting needles

Bent, 50 mm, 0,6 mm
Catalog # 631-7153
Supplier:   HAMMACHER KARL
Dissecting needles
Catalog # 631-7153
Supplier:  HAMMACHER KARL
Dissecting needles
Dissecting needles

Bent, 50 mm, 0,6 mm

Price
44,50 €
Log in for availability
1 * 10 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic.

Specifications
  • Ø:
    0,6 mm
  • Form:
    Bent
  • Length:
    50 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.