Orders LinkContactUsLinkComponent
Forceps, straight
Forceps, straight
Straight, fine tips, 160 mm
Catalog # 232-1918
Supplier:   HAMMACHER KARL
Forceps, straight
Catalog # 232-1918
Supplier:  HAMMACHER KARL
Forceps, straight
Forceps, straight
Straight, fine tips, 160 mm
Price
78,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic.
  • Smooth points
Specifications
  • Type:
    Straight, fine tips
  • Length:
    160 mm