Forceps, straight
Very delicate tips, straight, No. 4, 105 mm
Catalog # 232-2107
Supplier: HAMMACHER KARL
Jump to:
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic.
- Smooth points
Specifications
- Type:Very delicate tips, straight, No. 4
- Length:105 mm