Orders LinkContactUsLinkComponent
Forceps, straight
Forceps, straight
With fine cutting tips, 105 mm
Catalog # 232-0069
Supplier:   HAMMACHER KARL
Forceps, straight
Catalog # 232-0069
Supplier:  HAMMACHER KARL
Forceps, straight
Forceps, straight
With fine cutting tips, 105 mm
Price
142,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic.
  • Smooth points
Specifications
  • Type:
    With fine cutting tips
  • Length:
    105 mm