Orders LinkContactUsLinkComponent
Physiology scissors, straight
Physiology scissors, straight
Sharp/sharp, 145 mm
Catalog # 233-0086
Supplier:   HAMMACHER KARL
Physiology scissors, straight
Catalog # 233-0086
Supplier:  HAMMACHER KARL
Physiology scissors, straight
Physiology scissors, straight
Sharp/sharp, 145 mm
Price
127,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
  • Sharp-sharp
  • Ideal for a wide range of laboratory applications
  • Acid-resistant
  • Autoclavable
Specifications
  • Type:
    Sharp/sharp
  • Length:
    145 mm