Orders LinkContactUsLinkComponent
Thread and vessel scissors
Thread and vessel scissors
Straight, sharp/blunt, 110 mm
Catalog # 233-1117
Supplier:   HAMMACHER KARL
Thread and vessel scissors
Catalog # 233-1117
Supplier:  HAMMACHER KARL
Thread and vessel scissors
Thread and vessel scissors
Straight, sharp/blunt, 110 mm
Price
105,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
  • Sharp-blunt
  • Ideal for a wide range of laboratory applications
  • Acid-resistant
  • Autoclavable
Specifications
  • Type:
    Straight, sharp/blunt
  • Length:
    110 mm