Orders LinkContactUsLinkComponent
Dissecting scissors
Dissecting scissors
Slightly curved, sharp/sharp, 110 mm
Catalog # 233-1118
Supplier:   HAMMACHER KARL
Dissecting scissors
Catalog # 233-1118
Supplier:  HAMMACHER KARL
Dissecting scissors
Dissecting scissors
Slightly curved, sharp/sharp, 110 mm
Price
108,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
  • Ideal for a wide range of laboratory applications
  • Acid-resistant
  • Autoclavable
  • Sharp-sharp
Specifications
  • Type:
    Slightly curved, sharp/sharp
  • Length:
    110 mm