Order Entry
Belgium
Orders LinkContactUsLinkComponent
Spoon spatulas, micro

Spoon spatulas, micro

150 mm, 5×7 mm
Catalog # HAMMHSN061-15
Supplier:   HAMMACHER KARL
Spoon spatulas, micro
Catalog # HAMMHSN061-15
Supplier:  HAMMACHER KARL
Spoon spatulas, micro
Spoon spatulas, micro

150 mm, 5×7 mm

Price
12,50 €
Log in for availability
1 * 1 ST
Product Details & Documents

Double-ended spatulas with one spoon end and one spatula end. Made of stainless steel.

Specifications
  • Length:
    150 mm
  • Seq:
    02
  • Spoon size:
    5×7 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.