Orders LinkContactUsLinkComponent
Forceps
Forceps
London-College type, bent, pointed serrated tips, 150 mm
Catalog # HAMML-HSC020-05
Supplier:   HAMMACHER KARL
Forceps
Catalog # HAMML-HSC020-05
Supplier:  HAMMACHER KARL
Forceps
Forceps
London-College type, bent, pointed serrated tips, 150 mm
Price
25,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
Specifications
  • Type:
    London-College type, bent, pointed serrated tips
  • Length:
    150 mm