Orders LinkContactUsLinkComponent
Universal scissors, straight
Universal scissors, straight
Sharp-blunt, 200 mm
Catalog # 233-1154
Supplier:   HAMMACHER KARL
Universal scissors, straight
Catalog # 233-1154
Supplier:  HAMMACHER KARL
Universal scissors, straight
Universal scissors, straight
Sharp-blunt, 200 mm
Price
182,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel or chromium-plated steel.
  • Sharp-blunt
  • For a wide range of applications
  • Corrosion resistant
  • Easy action
Specifications
  • Type:
    Sharp-blunt
  • Length:
    200 mm
  • Seq:
    02