Orders LinkContactUsLinkComponent
Laboratory scissors, straight
Laboratory scissors, straight
Sharp/blunt, 180 mm
Catalog # 233-1114
Supplier:   HAMMACHER KARL
Laboratory scissors, straight
Catalog # 233-1114
Supplier:  HAMMACHER KARL
Laboratory scissors, straight
Laboratory scissors, straight
Sharp/blunt, 180 mm
Price
42,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel, with plastic handles.
  • Ideal for a wide range of laboratory applications
  • Corrosion resistant
  • Easy action
Specifications
  • Type:
    Sharp/blunt
  • Length:
    180 mm
  • Seq:
    03