Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Dissecting scissors
Dissecting scissors
Straight, sharp/sharp, 115 mm
Catalog # HAMML-HSB300-11
Supplier:   HAMMACHER KARL
Dissecting scissors
Catalog # HAMML-HSB300-11
Supplier:  HAMMACHER KARL
Dissecting scissors
Dissecting scissors
Straight, sharp/sharp, 115 mm
Price
286,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These dissecting scissors made of stainless steel feature tungsten carbide inserts.
Specifications
  • Type:
    Straight, sharp/sharp
  • Longueur:
    115 mm
  • Seq:
    01