Orders LinkContactUsLinkComponent
Operating scissors
Operating scissors
Curved, sharp/blunt, 165 mm
Catalog # 233-1241
Supplier:   HAMMACHER KARL
Operating scissors
Catalog # 233-1241
Supplier:  HAMMACHER KARL
Operating scissors
Operating scissors
Curved, sharp/blunt, 165 mm
Price
95,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
These operating scissors made from stainless steel are suitable for medical and clinical applications.
Specifications
  • Type:
    Curved, sharp/blunt
  • Length:
    165 mm
  • Seq:
    12