Orders LinkContactUsLinkComponent
Microscopy scissors
Microscopy scissors
Straight, sharp/sharp, with angular handles, 105 mm
Catalog # HAMML-HSB010-10
Supplier:   HAMMACHER KARL
Microscopy scissors
Catalog # HAMML-HSB010-10
Supplier:  HAMMACHER KARL
Microscopy scissors
Microscopy scissors
Straight, sharp/sharp, with angular handles, 105 mm
Price
64,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
These sharp microscopy scissors with angular handles are made from stainless steel.
Specifications
  • Type:
    Straight, sharp/sharp, with angular handles
  • Length:
    105 mm
  • Seq:
    01