Order Entry
Belgium
Orders LinkContactUsLinkComponent
Electrolyte Solutions

Electrolyte Solutions

Electrolyte Protelyte, 100 ml, 100 ml
Catalog # 662-9204
Supplier:   HAMILTON BONADUZ
Electrolyte Solutions
Catalog # 662-9204
Supplier:  HAMILTON BONADUZ
CAS Number:  
Electrolyte Solutions
Electrolyte Solutions

Electrolyte Protelyte, 100 ml, 100 ml

CAS Number:  
Price
40,10 €
Log in for availability
1 * 100 mL
Product Details & Documents

A variety of electrolyte solutions for pH and ORP electrodes.

Documents

Specifications

Specifications

  • Pk:
    100 ml
  • Description:
    Electrolyte Protelyte, 100 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.