Electrolyte Solutions
Electrolyte Skylyte, 100 ml, 100 ml
Catalog # 662-9203
Supplier: HAMILTON BONADUZ
Jump to:
Product Details & Documents
A variety of electrolyte solutions for pH and ORP electrodes.
Specifications
Specifications
- Pk:100 ml
- Description:Electrolyte Skylyte, 100 ml