Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Pestles

48 mm, 185 mm, 410-9113
Catalog # 410-9323
Pestles
Catalog # 410-9323
Pestles
Pestles

48 mm, 185 mm, 410-9113

Price
58,90 €
Log in for availability
1 * 1 ST
Product Details & Documents

Porcelain with glazed handle, glazed or unglazed grinding surface.

Certifications: Manufactured to DIN 12906.

Specifications
  • For:
    410-9113
  • Head Ø:
    48 mm
  • Length:
    185 mm
  • Seq:
    18
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.