Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pestles

Pestles

45 mm, 180 mm, 410-9327
Catalog # 410-9324
Pestles
Catalog # 410-9324
Pestles
Pestles

45 mm, 180 mm, 410-9327

Price
53,20 €
Log in for availability
1 * 1 ST
Product Details & Documents

Porcelain with glazed handle, glazed or unglazed grinding surface.

Certifications: Manufactured to DIN 12906.

Specifications
  • For:
    410-9327
  • Head Ø:
    45 mm
  • Length:
    180 mm
  • Seq:
    17
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.