Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Pestles

30 mm, 135 mm, 410-9328
  410-9132
Pestles
  410-9132
Pestles
Pestles

30 mm, 135 mm, 410-9328

 
23,90 €
   
1 * 1 ST
 

Porcelain with glazed handle, glazed or unglazed grinding surface.

 : Manufactured to DIN 12906.

 
  • For:
    410-9328
  • Head Ø:
    30 mm
  • Length:
    135 mm
  • Seq:
    14