Order Entry
Belgium
Orders LinkContactUsLinkComponent
Büchner funnels

Büchner funnels

10600 ml, 320 mm
Catalog # 511-2513
Büchner funnels
Catalog # 511-2513
Büchner funnels
Büchner funnels

10600 ml, 320 mm

Price
1 990,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

White glazed porcelain.

For filter paper or membrane filtration discs.

Certifications: DIN 12 905 (511-2502, 511-2501 and 511-2500 not according to DIN).

Specifications
  • Capacity:
    10600 ml
  • Seq:
    14
  • For filter Ø:
    320 mm