Order Entry
Belgium
Orders LinkContactUsLinkComponent
Iron standard solution

Iron standard solution

Iron Standard Solution, 100 ml (NIST), 100 mg/l as Fe, 100 mL
Catalog # HACH1417542
Supplier:   Hach
Iron standard solution
Catalog # HACH1417542
Supplier:  Hach
CAS Number:  
Iron standard solution
Iron standard solution

Iron Standard Solution, 100 ml (NIST), 100 mg/l as Fe, 100 mL

CAS Number:  
Price
27,90 €
Log in for availability
1 * 100 mL
Product Details & Documents

For accuracy checks in iron determinations.

  • Traceable to NIST
Specifications

Specifications

  • Concentration:
    100 mg/l as Fe
  • Pk:
    100 mL
  • Description:
    Iron Standard Solution, 100 ml (NIST)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.