Saccharometers
10 - 20%, 0,1, 4…26 °C
Catalog # 641-0002
Supplier: GERING
Jump to:
Product Details & Documents
These saccharometers with temperature correction scale are suitable for the official testing in breweries especially on wort.
Specifications
- Temperature Range:4…26 °C
- Seq:04
- Division:0,1
- Range:10 - 20%
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



