Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Brix saccharimeters

50 - 60 °Brix, 0,1, 350 mm
  614-0078
 :   GERING
Brix saccharimeters
  614-0078
 :  GERING
Brix saccharimeters
Brix saccharimeters

50 - 60 °Brix, 0,1, 350 mm

 
136,00 €
   
1 * 1 ST
 

Brix saccharimeters with a reference temperature of 20 °C.

 
  • Length:
    350 mm
  • Seq:
    16
  • Division:
    0,1
  • Range:
    50 - 60 °Brix